Assay Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136074.2 | NP_001129546.1 | 1 - 2 | 60 | 87 | |
| GenBank mRNA | AK044316.1 | - | 1 - 2 | 43 | 87 | |
| BC031440.1 | - | 1 - 2 | 44 | 4527590 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044316.1, BC031440.1, NM_001136074.2 transcripts.
Set Membership: |
|

Nrl
Genomic Map