Assay Details
| Gene Symbol: Pck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028994.2 | NP_083270.1 | 5 - 6 | 1066 | 68 | |
| GenBank mRNA | AK007429.1 | - | 2 - 3 | 260 | 68 | |
| AK030501.1 | - | 5 - 6 | 1067 | 68 | ||
| AK031612.1 | - | 5 - 6 | 911 | 68 | ||
| AK034927.1 | - | 5 - 6 | 945 | 68 | ||
| AK147972.1 | - | 5 - 6 | 920 | 68 | ||
| AK165094.1 | - | 5 - 6 | 944 | 68 | ||
| BC010318.1 | - | 5 - 6 | 929 | 3596227 | 68 | |
| BC023437.1 | - | 3 - 4 | 534 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pck2
Genomic Map