Assay Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145705.3 | NP_663751.2 | 2 - 3 | 518 | 60 | |
| GenBank mRNA | AF214013.1 | - | 2 - 3 | 509 | 60 | |
| AF518764.1 | - | 2 - 3 | 483 | 60 | ||
| AK011154.1 | - | 2 - 3 | 491 | 60 | ||
| AK016456.1 | - | 2 - 3 | 451 | 60 | ||
| AK028952.1 | - | 2 - 3 | 489 | 60 | ||
| AK090324.1 | - | 2 - 3 | 485 | 60 | ||
| AK144592.1 | - | 2 - 3 | 447 | 60 | ||
| AK186139.1 | - | 2 - 3 | 129 | 60 | ||
| BC030347.1 | - | 2 - 3 | 444 | 5003804 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tinf2
Genomic Map