Assay Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145705.3 | NP_663751.2 | 5 - 6 | 810 | 92 | |
| GenBank mRNA | AF214013.1 | - | 5 - 6 | 801 | 92 | |
| AF518764.1 | - | 5 - 6 | 775 | 92 | ||
| AK090324.1 | - | 5 - 6 | 777 | 92 | ||
| AK144592.1 | - | 5 - 6 | 739 | 92 | ||
| AK205058.1 | - | 1 - 2 | 92 | |||
| BC030347.1 | - | 5 - 6 | 736 | 5003804 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tinf2
Genomic Map