Assay Details
| Gene Symbol: Cideb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009894.3 | NP_034024.2 | 2 - 3 | 318 | 57 | |
| GenBank mRNA | AF041377.1 | - | 2 - 3 | 306 | 57 | |
| AK003968.1 | - | 2 - 3 | 355 | 57 | ||
| BC012664.1 | - | 2 - 3 | 282 | 4218866 | 57 | |
| BC046340.1 | - | 2 - 3 | 312 | 6478230 | 57 | |
| CB600615.1 | - | 2 - 3 | 295 | 57 | ||
More Information
Set Membership: |
|

Cideb
Genomic Map