Assay Details
Gene Symbol: Olfr746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146298.2 | NP_666410.2 | 1 | 896 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069679 | NM_146298.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50979 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069679 | NM_146298.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503407 | NM_146298.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597970 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597956 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597924 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597926 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597954 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597961 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597959 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597925 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29597962 | - | - |
Whole Mouse Genome G4122A | A_51_P503407 | NM_146298.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117324 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11148781 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11199676 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11199672 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11148779 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11199677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597956 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29597961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597952 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597967 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597935 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597925 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597962 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597927 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597942 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11696384 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597943 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597926 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11199664 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597981 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597934 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597954 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11199660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597955 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597950 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11148782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11256719 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29597940 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11433776 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10414492 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10414491 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 710553 | - | - |
ABI Mouse Genome Survey Microarray | 861006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 4480025 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004480025 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770327 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2770327 | - | - |
mouse-6 v1.1 (Illumina) | scl0258295.1_49-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4480025 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258295.1_49-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480025 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2780656 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2780656 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770327 | - | - |
