Assay Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134078.4 | NP_598839.2 | 7 - 8 | 1139 | 73 | |
| GenBank mRNA | BB621791.1 | - | 3 - 4 | 272 | 73 | |
| BC024115.1 | - | 7 - 8 | 1127 | 5100379 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chmp7
Genomic Map