Assay Details
| Gene Symbol: Phyhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145981.3 | NP_666093.1 | 3 - 4 | 643 | 89 | |
| GenBank mRNA | AK049900.1 | - | 3 - 4 | 644 | 89 | |
| BC030494.1 | - | 3 - 4 | 640 | 89 | ||
| BC139360.1 | - | 4 - 5 | 626 | 8862382 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phyhip
Genomic Map