Assay Details
| Gene Symbol: Chml | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021350.2 | NP_067325.2 | 1 | 1642 | 104 | |
| GenBank mRNA | BC118932.2 | - | 1 | 1642 | 40090670 | 104 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chml
Genomic Map