Assay Details
| Gene Symbol: Exo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012012.4 | NP_036142.2 | 1 - 2 | 75 | 67 | |
| GenBank mRNA | AK028728.1 | - | 1 - 2 | 56 | 67 | |
| BC006671.1 | - | 1 - 2 | 39 | 3499406 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028728.1, BC006671.1, NM_012012.4 transcripts.
Set Membership: |
|

Exo1
Genomic Map