| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100013780.mm.1 |
NM_012012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013780.mm.1 |
NM_012012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013780.mm.1 |
TC0100001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013778.mm.1 |
NM_012012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013778.mm.1 |
NM_012012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013778.mm.1 |
TC0100001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013781.mm.1 |
NM_012012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013781.mm.1 |
NM_012012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013781.mm.1 |
TC0100001656.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100013779.mm.1 |
NM_012012.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013779.mm.1 |
NM_012012.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100013779.mm.1 |
TC0100001656.mm.1 |
MTA TC |