Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300011353.mm.1 |
NM_009626.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011353.mm.1 |
NM_009626.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011353.mm.1 |
TC0300001408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011352.mm.1 |
NM_009626.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011352.mm.1 |
NM_009626.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011352.mm.1 |
TC0300001408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011350.mm.1 |
NM_009626.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011350.mm.1 |
NM_009626.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011350.mm.1 |
TC0300001408.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300011354.mm.1 |
NM_009626.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011354.mm.1 |
NM_009626.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300011354.mm.1 |
TC0300001408.mm.1 |
MTA TC |