Assay Details
| Gene Symbol: Adh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026945.1 | NP_081221.1 | 3 - 4 | 321 | 84 | |
| GenBank mRNA | AK007397.1 | - | 3 - 4 | 317 | 84 | |
| AK028114.1 | - | 3 - 4 | 322 | 84 | ||
| BC130016.1 | - | 3 - 4 | 290 | 40140468 | 84 | |
More Information
Set Membership: |
|

Adh6a
Genomic Map