Assay Details
| Gene Symbol: Dleu2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF380425.1 | - | LIT3421 DLEU2 |
1 - 2 | 165 | 141 | |
| AK029956.1 | - | 1 - 2 | 191 | 141 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 15a | Mir15a |
Mirn15a, mir-15a, mmu-mir-15a |
-11.3kb | Sense | Chr. 14 - 61632027 - 61632110 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 27017 |
Gm27017 | 31.3kb | Anti-sense | Chr. 14 - 61680039 - 61686642 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Dleu2
Genomic Map