Assay Details
| Gene Symbol: Klc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286038.1 | NP_001272967.1 | 4 - 5 | 782 | 87 | |
| NM_001286039.1 | NP_001272968.1 | 4 - 5 | 658 | 87 | ||
| NM_146182.4 | NP_666294.1 | 4 - 5 | 687 | 87 | ||
| GenBank mRNA | AK158094.1 | - | 4 - 5 | 624 | 87 | |
| BC017147.1 | - | 4 - 5 | 624 | 4506646 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klc3
Genomic Map