Assay Details
| Gene Symbol: Rsph6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031255.2 | NP_112545.2 | 3 - 4 | 1682 | 60 | |
| GenBank mRNA | AF329192.1 | - | 3 - 4 | 1675 | 60 | |
| AK029699.1 | - | 3 - 4 | 1682 | 60 | ||
| BC137992.1 | - | 3 - 4 | 1675 | 8861011 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rsph6a
Genomic Map