Assay Details
| Gene Symbol: Degs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007853.4 | NP_031879.1 | 1 - 2 | 137 | 101 | |
| GenBank mRNA | AF466376.1 | - | 1 - 2 | 154 | 101 | |
| AK002617.1 | - | 1 - 2 | 145 | 101 | ||
| AK077257.1 | - | 1 - 2 | 138 | 101 | ||
| AK149811.1 | - | 1 - 2 | 127 | 101 | ||
| AK150908.1 | - | 1 - 2 | 127 | 101 | ||
| AK163404.1 | - | 1 - 2 | 127 | 101 | ||
| AK165352.1 | - | 1 - 2 | 152 | 101 | ||
| BC003751.1 | - | 1 - 2 | 155 | 3600571 | 101 | |
| Y08460.1 | - | 1 - 2 | 122 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Degs1
Genomic Map