Assay Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009794.3 | NP_033924.2 | 12 - 13 | 1651 | 70 | |
| GenBank mRNA | AF015038.1 | - | 12 - 13 | 1597 | 70 | |
| AK052141.1 | - | 10 - 11 | 1239 | 70 | ||
| AK150695.1 | - | 12 - 13 | 1625 | 70 | ||
| AK152094.1 | - | 12 - 13 | 1625 | 70 | ||
| AK152790.1 | - | 12 - 13 | 1628 | 70 | ||
| AK152994.1 | - | 12 - 13 | 1628 | 70 | ||
| AK154855.1 | - | 12 - 13 | 1665 | 70 | ||
| AK183150.1 | - | 1 - 2 | 87 | 70 | ||
| AK183766.1 | - | 1 - 2 | 71 | |||
| AK199295.1 | - | 1 - 2 | 87 | 70 | ||
| BC054726.1 | - | 12 - 13 | 1651 | 5703563 | 70 | |
| D38117.1 | - | 12 - 13 | 1692 | 70 | ||
| Y10139.1 | - | 12 - 13 | 1533 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn2
Genomic Map