Assay Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009794.3 | NP_033924.2 | 16 - 17 | 1880 | 76 | |
| GenBank mRNA | AF015038.1 | - | 16 - 17 | 1826 | 76 | |
| AK052141.1 | - | 14 - 15 | 1468 | 76 | ||
| AK150695.1 | - | 16 - 17 | 1854 | 76 | ||
| AK152094.1 | - | 16 - 17 | 1854 | 76 | ||
| AK152790.1 | - | 16 - 17 | 1857 | 76 | ||
| AK152994.1 | - | 16 - 17 | 1857 | 76 | ||
| AK154855.1 | - | 16 - 17 | 1894 | 76 | ||
| AK183766.1 | - | 5 - 6 | 280 | 76 | ||
| AK195491.1 | - | 5 - 6 | 324 | 76 | ||
| BC054726.1 | - | 16 - 17 | 1880 | 5703563 | 76 | |
| D38117.1 | - | 16 - 17 | 1921 | 76 | ||
| Y10139.1 | - | 16 - 17 | 1762 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn2
Genomic Map