Assay Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009794.3 | NP_033924.2 | 6 - 7 | 942 | 73 | |
| GenBank mRNA | AF015038.1 | - | 6 - 7 | 888 | 73 | |
| AK052141.1 | - | 6 - 7 | 917 | 73 | ||
| AK053832.1 | - | 6 - 7 | 2228 | 73 | ||
| AK078787.1 | - | 2 - 3 | 1571 | 73 | ||
| AK150695.1 | - | 6 - 7 | 916 | 73 | ||
| AK152094.1 | - | 6 - 7 | 916 | 73 | ||
| AK154855.1 | - | 6 - 7 | 956 | 73 | ||
| BC054726.1 | - | 6 - 7 | 942 | 5703563 | 73 | |
| D38117.1 | - | 6 - 7 | 983 | 73 | ||
| Y10139.1 | - | 6 - 7 | 824 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn2
Genomic Map