Assay Details
| Gene Symbol: Trp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173378.2 | NP_775554.2 | 12 - 13 | 2206 | 86 | |
| GenBank mRNA | AK134556.1 | - | 5 - 6 | 1176 | 86 | |
| BC030894.1 | - | 4 - 5 | 835 | 86 | ||
| BC042874.1 | - | 11 - 12 | 1808 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp2
Genomic Map