Assay Details
| Gene Symbol: Trp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173378.2 | NP_775554.2 | 6 - 7 | 907 | 65 | |
| GenBank mRNA | BC042874.1 | - | 5 - 6 | 509 | 65 | |
| CN529251.1 | - | 2 - 3 | 195 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp2
Genomic Map