Assay Details
| Gene Symbol: Trp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173378.2 | NP_775554.2 | 9 - 10 | 1484 | 114 | |
| GenBank mRNA | AK134556.1 | - | 2 - 3 | 454 | 114 | |
| BC030894.1 | - | 1 - 2 | 113 | 114 | ||
| BC042874.1 | - | 8 - 9 | 1086 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trp53bp2
Genomic Map