Assay Details
| Gene Symbol: Rrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009102.3 | NP_033128.1 | 3 - 4 | 421 | 74 | |
| GenBank mRNA | AF012271.1 | - | 3 - 4 | 425 | 74 | |
| AK044717.1 | - | 409 | 74 | |||
| BC046288.1 | - | 4 - 5 | 547 | 6468540 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrh
Genomic Map