Assay Details
| Gene Symbol: Xpnpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289729.1 | NP_001276658.1 | 16 - 17 | 2115 | 67 | |
| NM_133213.2 | NP_573476.2 | 16 - 17 | 1914 | 67 | ||
| GenBank mRNA | AF367247.1 | - | 16 - 17 | 1578 | 67 | |
| AF428102.1 | - | 16 - 17 | 1765 | 67 | ||
| AK078835.1 | - | 16 - 17 | 1721 | 67 | ||
| AK085520.1 | - | 16 - 17 | 1913 | 67 | ||
| AK087627.1 | - | 16 - 17 | 1721 | 67 | ||
| AK164538.1 | - | 16 - 17 | 1742 | 67 | ||
| AK172086.1 | - | 16 - 17 | 1915 | 67 | ||
| BC140977.1 | - | 16 - 17 | 1558 | 9055589 | 67 | |
More Information
Set Membership: |
|

Xpnpep2
Genomic Map