Assay Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178782.4 | NP_848897.3 | 13 - 14 | 5154 | 60 | |
| GenBank mRNA | AK054259.1 | - | 9 - 10 | 1688 | 60 | |
| AK081004.1 | - | 9 - 10 | 3722 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcorl1
Genomic Map