| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00002892.mm.1 |
NM_178782.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002892.mm.1 |
NM_178782.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002892.mm.1 |
TC0X00000433.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002893.mm.1 |
NM_178782.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002893.mm.1 |
NM_178782.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002893.mm.1 |
TC0X00000433.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002889.mm.1 |
NM_178782.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002889.mm.1 |
NM_178782.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002889.mm.1 |
TC0X00000433.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002895.mm.1 |
NM_178782.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002895.mm.1 |
NM_178782.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002895.mm.1 |
TC0X00000433.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002887.mm.1 |
NM_178782.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002887.mm.1 |
NM_178782.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002887.mm.1 |
TC0X00000433.mm.1 |
MTA TC |