Assay Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178782.4 | NP_848897.3 | 6 - 7 | 3863 | 67 | |
| GenBank mRNA | AK054259.1 | - | 2 - 3 | 397 | 67 | |
| AK081004.1 | - | 2 - 3 | 2431 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcorl1
Genomic Map