Assay Details
| Gene Symbol: Utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028276.1 | NP_082552.1 | 1 - 2 | 39 | 110 | |
| GenBank mRNA | AK012492.1 | - | 1 - 2 | 79 | 110 | |
| AK134348.1 | - | 1 - 2 | 97 | 110 | ||
| AK134784.1 | - | 1 - 2 | 64 | 110 | ||
| AK165575.1 | - | 1 - 2 | 79 | 110 | ||
| AK165628.1 | - | 1 - 2 | 79 | 110 | ||
| AK172924.1 | - | 1 - 2 | 39 | 110 | ||
| AY316163.1 | - | 1 - 2 | 62 | 110 | ||
| BC062165.1 | - | 1 - 2 | 88 | 110 | ||
| BC082590.1 | - | 1 - 2 | 65 | 30625504 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012492.1, AK134348.1, AK134784.1, AY316163.1, BC062165.1, BC082590.1, NM_028276.1 transcripts.
Set Membership: |
|

Utp14a
Genomic Map