| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00002863.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002863.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002863.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002866.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002866.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002866.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002865.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002865.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002865.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002864.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002864.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002864.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002867.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002867.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002867.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002868.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002868.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002868.mm.1 |
TC0X00000429.mm.1 |
MTA TC |