| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00002872.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002872.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002872.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002874.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002874.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002874.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002876.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002876.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002876.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002873.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002873.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002873.mm.1 |
TC0X00000429.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002875.mm.1 |
NM_028276.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002875.mm.1 |
NM_028276.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002875.mm.1 |
TC0X00000429.mm.1 |
MTA TC |