Assay Details
Gene Symbol: Rbm48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172991.4 | NP_766579.3 | 4 - 5 | 1498 | 72 | |
GenBank mRNA | AK047811.1 | - | 4 - 5 | 1030 | 72 | |
BC029549.1 | - | 4 - 5 | 1362 | 5012510 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445967 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445970 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445968 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445969 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445966 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | AK041876.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | AK047811.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | BC029549.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | NM_172991.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | AK041876.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | AK047811.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | BC029549.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P475450 | NM_172991.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582555 | AK047811.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582555 | BC029549.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582555 | NM_172991.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582555 | AK047811.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582555 | BC029549.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582555 | NM_172991.4 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740841 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136263 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274596 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379398 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521138 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112202 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136231 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133406 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582555 | AK047811.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582555 | BC029549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582555 | NM_172991.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582555 | AK047811.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582555 | BC029549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582555 | NM_172991.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | AK041876.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | AK047811.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | BC029549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | NM_172991.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | AK041876.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | AK047811.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | BC029549.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P475450 | NM_172991.4 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445978 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23446001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23445975 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133406 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136231 | - | - |
Whole Mouse Genome G4122A | A_52_P475454 | AK041876.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475454 | AK047811.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475454 | BC029549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475454 | NM_172991.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | AK041876.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | AK047811.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | BC029549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | NM_172991.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | AK041876.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | AK047811.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | BC029549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P475450 | NM_172991.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P582555 | AK047811.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P582555 | BC029549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P582555 | NM_172991.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P582555 | AK047811.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P582555 | BC029549.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P582555 | NM_172991.4 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128376 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124479 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136231 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133406 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04606887 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04606890 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04606886 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04606892 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04606888 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445967 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445968 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445966 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445969 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23445970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445972 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23446010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23445969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04827553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04606888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04185602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04817892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04817890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04937017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04937019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04579584 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04185600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04606884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04631658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23445999 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23446003 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10527884 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519353 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10527881 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10527885 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10527882 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10527883 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1433957_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10527880 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10519354 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10519346 | - | - |
Mouse Genome 430 2.0 Array | 1433957_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1433957_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269623_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269623_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500017339.mm.1 | TC0500001885.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500017339.mm.1 | NM_172991.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500017339.mm.1 | NM_172991.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500017338.mm.1 | TC0500001885.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500017338.mm.1 | NM_172991.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500017338.mm.1 | NM_172991.4 | Refseq |
Mouse Expression Set 430 | 1433957_at | - | - |
Mouse Expression Set 430 | 1433957_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433957_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433957_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 635033 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 103170577 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 670019 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000670019 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685436 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2570462 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2685436 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225045 | - | - |
mouse-6 v1.1 (Illumina) | scl28121.7_60-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 670019 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103170577 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl28121.7_60-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670019 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103170577 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2940358 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1259185 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2570461 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259185 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2940358 | - | - |
