Assay Details
| Gene Symbol: Fam122b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166365.2 | NP_001159837.2 | 3 - 4 | 1054 | 76 | |
| NM_001166583.1 | NP_001160055.1 | 3 - 4 | 1054 | 76 | ||
| NM_030167.4 | NP_084443.1 | 3 - 4 | 1054 | 76 | ||
| GenBank mRNA | AB541013.1 | - | 3 - 4 | 222 | 76 | |
| AK019480.1 | - | 3 - 4 | 281 | 76 | ||
| AK036417.1 | - | 3 - 4 | 1051 | 76 | ||
| BC066167.1 | - | 3 - 4 | 269 | 30433483 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam122b
Genomic Map