Assay Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008150.2 | NP_032176.2 | 5 - 6 | 1157 | 137 | |
| GenBank mRNA | BC006622.1 | - | 5 - 6 | 1157 | 3967797 | 137 |
| CF168390.1 | - | 2 - 3 | 180 | 137 | ||
| X83577.1 | - | 5 - 6 | 1478 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpc4
Genomic Map