Assay Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008150.2 | NP_032176.2 | 7 - 8 | 1438 | 63 | |
| GenBank mRNA | AI892739.1 | - | 1 - 2 | 137 | 63 | |
| BC006622.1 | - | 7 - 8 | 1438 | 3967797 | 63 | |
| CF168390.1 | - | 4 - 5 | 461 | 63 | ||
| X83577.1 | - | 7 - 8 | 1759 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpc4
Genomic Map