Assay Details
| Gene Symbol: Rhcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019799.3 | NP_062773.2 | 1 - 2 | 314 | 71 | |
| GenBank mRNA | AF193810.1 | - | 1 - 2 | 305 | 71 | |
| AK076757.1 | - | 1 - 2 | 314 | 71 | ||
| AY254686.1 | - | 1 - 2 | 189 | 71 | ||
| BC119045.1 | - | 1 - 2 | 273 | 8733794 | 71 | |
| BC137652.1 | - | 1 - 2 | 255 | 8860671 | 71 | |
More Information
Set Membership: |
|

Rhcg
Genomic Map