Assay Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017462.2 | NP_059490.2 | 15 - 16 | 2777 | 89 | |
| GenBank mRNA | AB121698.1 | - | 14 - 15 | 2470 | 89 | |
| AK133584.1 | - | 15 - 16 | 2703 | 89 | ||
| AK148091.1 | - | 15 - 16 | 2739 | 89 | ||
| AK162462.1 | - | 1 - 2 | 269 | 89 | ||
| BC053024.1 | - | 15 - 16 | 2703 | 89 | ||
| BC061472.1 | - | 15 - 16 | 2691 | 6827238 | 89 | |
| BC080291.1 | - | 15 - 16 | 2608 | 89 | ||
| U53584.1 | - | 15 - 16 | 89 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Polg
Genomic Map