Assay Details
| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018811.6 | NP_061281.3 | 5 - 6 | 720 | 60 | |
| GenBank mRNA | AF201730.1 | - | 5 - 6 | 709 | 60 | |
| AF546701.1 | - | 5 - 6 | 721 | 60 | ||
| AK008690.1 | - | 5 - 6 | 730 | 60 | ||
| AK016296.1 | - | 3 - 4 | 381 | 60 | ||
| AK139526.1 | - | 5 - 6 | 729 | 60 | ||
| AK153642.1 | - | 5 - 6 | 767 | 60 | ||
| BC027098.1 | - | 5 - 6 | 690 | 5356667 | 60 | |
| BC089477.1 | - | 5 - 6 | 700 | 6741521 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd2
Genomic Map