Assay Details
| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018811.6 | NP_061281.3 | 8 - 9 | 1104 | 99 | |
| GenBank mRNA | AF201730.1 | - | 8 - 9 | 1093 | 99 | |
| AF546701.1 | - | 8 - 9 | 1105 | 99 | ||
| AK008690.1 | - | 8 - 9 | 1114 | 99 | ||
| AK016296.1 | - | 6 - 7 | 765 | 99 | ||
| AK139526.1 | - | 8 - 9 | 1113 | 99 | ||
| AK153642.1 | - | 8 - 9 | 1151 | 99 | ||
| BC027098.1 | - | 8 - 9 | 1074 | 5356667 | 99 | |
| BC089477.1 | - | 8 - 9 | 1084 | 6741521 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Abhd2
Genomic Map