Assay Details
| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161665.1 | NP_001155137.1 | 7 - 8 | 2090 | 66 | |
| GenBank mRNA | AB355846.1 | - | 7 - 8 | 2090 | 66 | |
| BC056349.1 | - | 2 - 3 | 173 | 6405419 | 66 | |
| BC068141.1 | - | 4 - 5 | 902 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC056349.1 transcripts.
Set Membership: |
|

Kif26b
Genomic Map