Assay Details
| Gene Symbol: Kif26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161665.1 | NP_001155137.1 | 11 - 12 | 2861 | 92 | |
| GenBank mRNA | AB355846.1 | - | 11 - 12 | 2861 | 92 | |
| BC056349.1 | - | 6 - 7 | 944 | 6405419 | 92 | |
| BC068141.1 | - | 8 - 9 | 1673 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif26b
Genomic Map