Assay Details
| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033285.1 | NP_001028457.1 | 28 - 29 | 3727 | 99 | |
| GenBank mRNA | BC158017.1 | - | 28 - 29 | 3727 | 9007323 | 99 |
| BC158095.1 | - | 27 - 28 | 3688 | 9007382 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc42bpa
Genomic Map