Assay Details
| Gene Symbol: Lgals3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145953.1 | NP_001139425.1 | 1 - 2 | 117 | 60 | |
| GenBank mRNA | AK149783.1 | - | 1 - 2 | 64 | 60 | |
| AK159536.1 | - | 1 - 2 | 58 | 60 | ||
| BC132328.1 | - | 1 - 2 | 43 | 40130605 | 60 | |
| BC138790.1 | - | 1 - 2 | 43 | 8861812 | 60 | |
| BC145419.1 | - | 1 - 2 | 43 | 9053977 | 60 | |
| X16834.1 | - | 1 - 2 | 36 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149783.1, AK159536.1, BC132328.1, BC138790.1, BC145419.1, NM_001145953.1, X16834.1 transcripts.
Set Membership: |
|

Lgals3
Genomic Map