Assay Details
| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144553.2 | NP_653136.2 | 12 - 13 | 1800 | 81 | |
| GenBank mRNA | AB076696.1 | - | 12 - 13 | 1800 | 81 | |
| AK036384.1 | - | 12 - 13 | 1602 | 81 | ||
| AK048470.1 | - | 12 - 13 | 1570 | 81 | ||
| AK085882.1 | - | 12 - 13 | 1581 | 81 | ||
| AK129032.1 | - | 12 - 13 | 1546 | 81 | ||
| AK135820.1 | - | 12 - 13 | 1607 | 81 | ||
| AK147955.1 | - | 12 - 13 | 1570 | 81 | ||
| BC027273.1 | - | 12 - 13 | 1521 | 81 | ||
| BC043924.1 | - | 4 - 5 | 426 | 81 | ||
| BC058087.1 | - | 12 - 13 | 1534 | 6816660 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap5
Genomic Map