Assay Details
| Gene Symbol: Dlgap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144553.2 | NP_653136.2 | 15 - 16 | 2298 | 146 | |
| GenBank mRNA | AB076696.1 | - | 15 - 16 | 2298 | 146 | |
| AK036384.1 | - | 14 - 15 | 3536 | 146 | ||
| AK048470.1 | - | 15 - 16 | 2069 | 146 | ||
| AK085882.1 | - | 15 - 16 | 2079 | 146 | ||
| AK129032.1 | - | 15 - 16 | 2044 | 146 | ||
| AK135820.1 | - | 15 - 16 | 2105 | 146 | ||
| AK147955.1 | - | 15 - 16 | 2068 | 146 | ||
| BC027273.1 | - | 15 - 16 | 2019 | 146 | ||
| BC043924.1 | - | 7 - 8 | 924 | 146 | ||
| BC058087.1 | - | 15 - 16 | 2032 | 6816660 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlgap5
Genomic Map