Assay Details
| Gene Symbol: Gch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008102.3 | NP_032128.1 | 1 - 2 | 455 | 83 | |
| GenBank mRNA | AK150657.1 | - | 1 - 2 | 467 | 83 | |
| AK152571.1 | - | 1 - 2 | 467 | 83 | ||
| BC005643.1 | - | 1 - 2 | 455 | 3709238 | 83 | |
| BC069921.1 | - | 1 - 2 | 459 | 6529016 | 83 | |
| L09737.1 | - | 1 - 2 | 442 | 83 | ||
| S71373.1 | - | 1 - 2 | 172 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gch1
Genomic Map