Assay Details
| Gene Symbol: Gch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008102.3 | NP_032128.1 | 2 - 3 | 559 | 122 | |
| GenBank mRNA | AK150657.1 | - | 2 - 3 | 571 | 122 | |
| AK152571.1 | - | 2 - 3 | 571 | 122 | ||
| BC005643.1 | - | 2 - 3 | 559 | 3709238 | 122 | |
| BC069921.1 | - | 2 - 3 | 563 | 6529016 | 122 | |
| L09737.1 | - | 2 - 3 | 546 | 122 | ||
| S71373.1 | - | 2 - 3 | 276 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gch1
Genomic Map