Assay Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172598.3 | NP_766186.2 | 15 - 16 | 2188 | 69 | |
| GenBank mRNA | AK036390.1 | - | 15 - 16 | 2258 | 69 | |
| BC063740.1 | - | 15 - 16 | 2121 | 6328626 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdhd1
Genomic Map