Assay Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172598.3 | NP_766186.2 | 21 - 22 | 2868 | 59 | |
| GenBank mRNA | AK036390.1 | - | 21 - 22 | 2938 | 59 | |
| BC063740.1 | - | 21 - 22 | 2801 | 6328626 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Wdhd1
Genomic Map