Assay Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172598.3 | NP_766186.2 | 22 - 23 | 3033 | 140 | |
| GenBank mRNA | AK160777.1 | - | 2 - 3 | 194 | 140 | |
| BC063740.1 | - | 22 - 23 | 2966 | 6328626 | 140 | |
More Information
Set Membership: |
|

Wdhd1
Genomic Map